3-Bromo-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15572-25G
76908-120EA 60.77 USD
76908-120
3-Bromo-5-fluorobenzaldehyde ≥97%
3-Bromo-5-fluorobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD04116319
CAS Number: 188813-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR