5-Methyl-2-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Danger

A512310-1G
76878-164EA 83.43 USD
76878-164
5-Methyl-2-(trifluoromethyl)benzonitrile 98%
5-Methyl-2-(trifluoromethyl)benzonitrile

Caution: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
MDL Number: MFCD06660290
CAS Number: 886502-61-0
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR