2-Tridecanone 95%

Supplier: Ambeed
Danger

A188790-25G A188790-5G
76825-182EA 7.21 USD
76825-182 76825-180
2-Tridecanone 95%
2-Tridecanone

Caution: for Research Use Only

Formula: C₁₃H₂₆O
MW: 198.35 g/mol
MDL Number: MFCD00008968
CAS Number: 593-08-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR