n-Dotriacontane 98%

Supplier: Ambeed
Warning

Synonyms: Dotriacontane

A822765-5G A822765-25G A822765-1G
76807-614EA 57.68 USD
76807-614 76807-616 76807-612
n-Dotriacontane 98%
n-Dotriacontane

Caution: for Research Use Only

Formula: H₃C(CH₂)₃₀CH₃
MW: 450.88 g/mol
MDL Number: MFCD00009411
CAS Number: 544-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR