1-Heneicosanol 97%

Supplier: Ambeed
Warning

Synonyms: HeneicosanolHeneicosyl alcohol

A712001-1MG A712001-100MG
76806-792EA 5.15 USD
76806-792 76806-790
1-Heneicosanol 97%
1-Heneicosanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₉CH₂OH
MW: 312.58 g/mol
MDL Number: MFCD00062834
CAS Number: 15594-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR