(5-Bromopentyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-phenylpentane

A589985-250MG
76805-890EA 30.9 USD
76805-890
(5-Bromopentyl)benzene 95%
(5-Bromopentyl)benzene

Caution: for Research Use Only

Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
MDL Number: MFCD01075177
CAS Number: 14469-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR