1,4-Dithioerythritol 98%

Supplier: Ambeed
Danger

Synonyms: Cleland's ReagentDL-Dithioerythritoltrans-1,4-Dimercaptobutane-2,3-diolDTEDithioerythritol(R*,S*)-1,4-Dimercaptobutane-2,3-diolErythro-1,4-Dimercaptobutane-2,3-diol

A423394-250MG A423394-5G A423394-1MG A423394-25G A423394-1G
76804-152EA 11.33 USD
76804-152 76804-150 76804-156 76804-154 76804-148
1,4-Dithioerythritol 98%
1,4-Dithioerythritol

Caution: for Research Use Only

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
MDL Number: MFCD00063750
CAS Number: 6892-68-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR