Ethyl-4-chloroquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

A223801-5G
76800-928EA 70.02 USD
76800-928
Ethyl-4-chloroquinoline-3-carboxylate 98%
Ethyl 4-chloroquinoline-3-carboxylate

Caution: for Research Use Only

Formula: C₁₂H₁₀ClNO₂
MW: 235.67 g/mol
MDL Number: MFCD00173407
CAS Number: 13720-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR