2'-Bromo-4',5'-(ethylenedioxy)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)ethanone

A124353-1G
76797-598EA 115.36 USD
76797-598
2'-Bromo-4',5'-(ethylenedioxy)acetophenone 98%
2'-Bromo-4',5'-(ethylenedioxy)acetophenone

Caution: for Research Use Only

Formula: C₁₀H₉BrO₃
MW: 257.0807 g/mol
MDL Number: MFCD00168681
CAS Number: 59820-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR