2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4-(4-Aminophenyl)piperazin-1-yl)ethanol
, 4-[4-(2-Hydroxyethyl)piperazin-1-yl]aniline
A190884-5G
A190884-250MG
A190884-1G
A190884-100MG
76784-902EA
202.91
USD
76784-902
76784-900
76784-906
76784-904
2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol 97%
2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol
Caution: for Research Use Only
Formula:
C₁₂H₁₉N₃O MW: 221.3 g/mol |
MDL Number:
MFCD00702247 CAS Number: 5521-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...