2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: AlbalonNaphazoline Hydrochloride

A792871-1MG A792871-25G A792871-100G
76728-478EA 12.36 USD
76728-478 76728-476 76728-474
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 98%
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride

Caution: for Research Use Only

Formula: C₁₄H₁₅ClN₂
MW: 246.738 g/mol
MDL Number: MFCD00012554
CAS Number: 550-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR