n-Heneicosane 98%

Supplier: Ambeed
Warning

A349290-5G A349290-25G A349290-1G
76724-430EA 248.23 USD
76724-430 76724-428 76724-432
n-Heneicosane 98%
n-Heneicosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
MDL Number: MFCD00009346
CAS Number: 629-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR