n-Eicosane 99+%

Supplier: Ambeed
Danger

Synonyms: N-Eicosane

A174110-25G
76720-624EA 16.48 USD
76720-624
n-Eicosane 99+%
n-Eicosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₈CH₃
MW: 282.55 g/mol
MDL Number: MFCD00009344
CAS Number: 112-95-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR