4-(Isobutyro)thioanisole 98%

Supplier: Ambeed
Warning

Synonyms: P-(methylthio)isobutyrophenone2-Methyl-1-[4-(methylsulphanyl)phenyl]propan-1-one

A151831-5G
76719-776EA 29.87 USD
76719-776
4-(Isobutyro)thioanisole 98%
4-(Isobutyro)thioanisole

Caution: for Research Use Only

Formula: C₁₁H₁₄OS
MW: 194.2933 g/mol
MDL Number: MFCD03094661
CAS Number: 53207-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR