N-Boc-S-methyl-L-cysteine 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Cys(Me)-OH
A526864-250MG
A526864-1G
A526864-25G
76695-886EA
42.23
USD
76695-886
76695-888
76695-890
N-Boc-S-methyl-L-cysteine 96%
N-Boc-S-methyl-L-cysteine
Caution: for Research Use Only
Formula:
C₉H₁₇NO₄S MW: 235.298 g/mol |
MDL Number:
MFCD00076923 CAS Number: 16947-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...