Diaminomaleonitrile 98%

Supplier: Ambeed
Danger

A303388-5G A303388-25G A303388-10G
76693-444EA 6.18 USD
76693-444 76693-442 76693-440
Diaminomaleonitrile 98%
Diaminomaleonitrile

Caution: for Research Use Only

Formula: C₄H₄N₄
MW: 108.1 g/mol
MDL Number: MFCD00001870
CAS Number: 1187-42-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR