1-Bromooctadecane 98%

Supplier: Ambeed
Warning

A1471064-500G A1471064-100G A1471064-25G
76662-774EA 32.96 USD
76662-774 76662-772 76662-770
1-Bromooctadecane 98%
1-Bromooctadecane

Caution: for Research Use Only

Formula: BrH₂C(CH₂)₁₆CH₃
MW: 333.4 g/mol
MDL Number: MFCD00000231
CAS Number: 112-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR