Sodium molybdate(VI) dihydrate ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium molybdenum oxide dihydrateMolybdic acid sodium salt dihydrateSodium molybdate dihydrateMolybdic acid, sodium salt dihydratedi-Sodium molybdate dihydrateMolybdic acid disodium salt dihydrate

44636-0250
TS44636-0250EA 46 USD
TS44636-0250
Sodium molybdate(VI) dihydrate ACS
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Density: 3.28 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6

Order Now

Specification Test Results

Appearance White Solid
Titration Complexometric 99.5 to 103.0 %
pH 7.0 to 10.5 (5% solution at 25.0 °C)
Chloride (Cl) ≤0.005 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.015 %
Ammonium (NH4) ≤0.001 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR