2-Nitro-5-(trifluoromethyl)benzenol

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Hydroxy-4-nitrobenzotrifluoride2-Nitro-5-(trifluoromethyl)phenol

PC6361-1G PC6361-5G
76603-664EA 120 USD
76603-664 76603-662
2-Nitro-5-(trifluoromethyl)benzenol
2-Nitro-5-(trifluoromethyl)benzenol
Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08282785
CAS Number: 402-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR