1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC9318-1G
76610-292EA 81 USD
76610-292
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid 97%
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid
Formula: C₁₂H₁₂FNO₄
MW: 253.23 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00202775
CAS Number: 497163-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR