5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5-Bromo-3-fluoropyridine-2-carboxylic acid

PC6074-5G PC6074-1G
76603-472EA 35 USD
76603-472 76603-474
5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%
5-Bromo-3-fluoro-2-pyridine carboxylic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR