1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Supplier: APOLLO SCIENTIFIC
Danger

PC6316-1G
76614-422EA 113 USD
76614-422
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
Formula: C₁₄H₁₃Cl₂F₃N₄S
MW: 397.25 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00172020
CAS Number: 303150-21-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR