1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(4-Chlorobenzoyl)-1,1,1-trifluoroacetone

PC4937-25G PC4937-10G PC4937-5G
76607-602EA 133 USD
76607-602 76605-874 76606-336
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Formula: C₁₀H₆ClF₃O₂
MW: 250.61 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00511276
CAS Number: 18931-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR