2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC

Danger

PC3728E-1G
76618-224EA 36 USD
76618-224
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile 97%
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile
Formula: C₁₄H₉ClFNO
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00068205
CAS Number: 175204-10-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR