4-(Trifluoromethyl)piperidine hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

PC7713-5G PC7713-1G
76608-750EA 52 USD
76608-750 76608-748
4-(Trifluoromethyl)piperidine hydrochloride
4-(Trifluoromethyl)piperidine hydrochloride
Formula: C₆H₁₁ClF₃N
Melting Pt: 155…160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD04971993
CAS Number: 155849-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR