Methyl-2,5-difluoro-4-nitrobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2,5-difluoro-4-nitrobenzoate

PC8637-1G PC8637-5G
76609-498EA 13 USD
76609-498 76609-500
Methyl-2,5-difluoro-4-nitrobenzoate
Methyl 2,5-difluoro-4-nitrobenzoate
Formula: C₈H₅F₂NO₄
MW: 217.13 g/mol
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD08689670
CAS Number: 924868-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR