2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC3823R-50MG PC3823R-1G PC3823R-5G
76626-420EA 19 USD
76626-420 76626-422 76626-418
2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%
2-Fluoro-6-(4-methylphenoxy)benzonitrile
Formula: C₁₄H₁₀FNO
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00068203
CAS Number: 175204-08-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR