2-Amino-4-fluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC

Warning

PC5484-25G PC5484-1G PC5484-5G
76627-062EA 13 USD
76627-062 76627-066 76627-064
2-Amino-4-fluorobenzonitrile 98%
2-Amino-4-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.1300 g/mol
Melting Pt: 110…111 °C
Storage Temperature: Ambient
MDL Number: MFCD09955451
CAS Number: 80517-22-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR