5-Fluoro-m-toluidine 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Fluoro-5-methylaniline

PC5923-5G PC5923-25G PC5923-1G
76603-344EA 80 USD
76603-344 76603-346
5-Fluoro-m-toluidine 97%
5-Fluoro-m-toluidine
Formula: C₇H₈FN
MW: 125.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077538
CAS Number: 52215-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR