2-(Difluoromethoxy)phenylacetic acid 98%

Supplier: APOLLO SCIENTIFIC

PC9251-1G
76610-240EA 47 USD
76610-240
2-(Difluoromethoxy)phenylacetic acid 98%
2-(Difluoromethoxy)phenylacetic acid
Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00236237
CAS Number: 86867-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR