Methyl-3,5-bis(trifluoromethyl)benzoate 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3,5-bis(trifluoromethyl)benzoate

PC5076-100G PC5076-5G PC5076-25G
76604-040EA 104 USD
76604-040 76607-048 76607-874
Methyl-3,5-bis(trifluoromethyl)benzoate 98%
Methyl 3,5-bis(trifluoromethyl)benzoate
Formula: C₁₀H₆F₆O₂
MW: 272.15 g/mol
Boiling Pt: 85…87 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052317
CAS Number: 26107-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR