2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile

Supplier: APOLLO SCIENTIFIC

Danger

PC31842-250MG PC31842-1G
76626-398EA 19 USD
76626-398 76626-396
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
Formula: C₁₄H₁₀FNOS
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00068069
CAS Number: 148901-52-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR