4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC

Warning

PC53378-10G PC53378-1G PC53378-5G
76601-148EA 373 USD
76601-148 76601-144 76601-146
4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde
Formula: C₇H₃BrFNO₃
MW: 248.0060 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10758067
CAS Number: 679839-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR