5-(3-(Trifluoromethyl)phenyl)oxazole

Supplier: APOLLO SCIENTIFIC

Warning

PC6225-1G PC6225-5G
76603-576EA 96 USD
76603-576 76603-574
5-(3-(Trifluoromethyl)phenyl)oxazole
5-(3-(Trifluoromethyl)phenyl)oxazole
Formula: C₁₀H₆F₃NO
MW: 213.1600 g/mol
Boiling Pt: 78 °C/0.6mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00085180
CAS Number: 175205-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR