5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole

Supplier: APOLLO SCIENTIFIC

Warning

PC8744-1G PC8744-5G PC8744-10G
76609-592EA 391 USD
76609-592 76609-590 76609-588
5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole
5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole
Formula: C₁₀H₇F₃N₂O
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD07787059
CAS Number: 82360-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR