5-Fluoro-3-iodo-1H-indazole 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC8399-5G PC8399-1G
76609-390EA 20 USD
76609-390 76609-392
5-Fluoro-3-iodo-1H-indazole 97%
5-Fluoro-3-iodo-1H-indazole
Formula: C₇H₄FIN₂
Storage Temperature: Ambient
MDL Number: MFCD06739145
CAS Number: 858629-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR