4-Bromo-6-fluoro-1H-indazole 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC908905-1G PC908905-250MG
76610-034EA 38 USD
76610-034 76610-032
4-Bromo-6-fluoro-1H-indazole 95+%
4-Bromo-6-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781448
CAS Number: 885520-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR