3-Amino-4-fluoro-1-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Amino-4-fluoro-1-methylindazole

PC5685-100MG PC5685-250MG
76602-668EA 55 USD
76602-668 76602-670
3-Amino-4-fluoro-1-methyl-1H-indazole
3-Amino-4-fluoro-1-methyl-1H-indazole
Formula: C₈H₈FN₃
MW: 165.1700 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD11109397
CAS Number: 162502-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR