Ethyl-3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

PC6154-1G PC6154-500MG
76603-510EA 62 USD
76603-510 76603-512
Ethyl-3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈BrF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12025928
CAS Number: 1160474-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR