4-Fluoro-3-iodobenzoic acid

Supplier: APOLLO SCIENTIFIC

Warning

PC9217-25G PC9217-5G PC9217-1G
76610-222EA 29 USD
76610-222 76610-226 76610-224
4-Fluoro-3-iodobenzoic acid
4-Fluoro-3-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD03788556
CAS Number: 403-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR