2-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC57059-1G
76602-976EA 152.8 USD
76602-976
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD16652441
CAS Number: 1240257-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR