4-(2-Fluorophenyl)-3-thiosemicarbazide 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: N-(2-Fluorophenyl)hydrazinecarbothioamide

PC4175J-1G PC4175J-2.5G
76618-310EA 14 USD
76618-310 76618-312
4-(2-Fluorophenyl)-3-thiosemicarbazide 97%
4-(2-Fluorophenyl)-3-thiosemicarbazide
Formula: C₇H₈FN₃S
MW: 185.22
Melting Pt: 154…157 °C (dec.)
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR