(2-Fluorophenyl)methanesulfonyl chloride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC4579-250MG
PC4579-1G
76623-612EA
22
USD
76623-612
76623-610
(2-Fluorophenyl)methanesulfonyl chloride 95%
(2-Fluorophenyl)methanesulphonyl chloride
Formula:
C₇H₆ClFO₂S Melting Pt: 54.5…56 °C Storage Temperature: Ambient |
UN:
3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...