3-Bromo-4-fluorobiphenyl 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-4-fluoro-1,1'-biphenyl

PC6677-1G
76618-860EA 25 USD
76618-860
3-Bromo-4-fluorobiphenyl 97%
3-Bromo-4-fluorobiphenyl
Formula: C₁₂H₈BrF
MW: 251.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01571091
CAS Number: 306935-88-6
UN: 3151
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR