2-Fluoro-4-iodobenzaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

PC9094-1G PC9094-25G PC9094-5G
76610-058EA 162 USD
76610-058 76610-052 76610-050
2-Fluoro-4-iodobenzaldehyde
2-Fluoro-4-iodobenzaldehyde
Formula: C₇H₄FIO
Storage Temperature: Ambient
MDL Number: MFCD07782056
CAS Number: 699016-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR