4-Fluoro-3-nitrophenol 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC49179-1G
PC49179-5G
PC49179-25G
76606-954EA
31
USD
76606-954
76607-108
76606-966
4-Fluoro-3-nitrophenol 97%
4-Fluoro-3-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.0993 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04038714 CAS Number: 2105-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...