3,5-Difluoro-2-nitrophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC9743-1G PC9743-5G PC9743-10G
76610-540EA 220 USD
76610-540 76610-544 76610-542
3,5-Difluoro-2-nitrophenol 98%
3,5-Difluoro-2-nitrophenol
Formula: C₆H₃F₂NO₃
MW: 175.0900 g/mol
Melting Pt: 46…47 °C
Storage Temperature: Ambient
MDL Number: MFCD08458165
CAS Number: 151414-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR