2-Chloro-5-fluoronicotinic acid 96%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC9179-25G
PC9179-5G
76610-192EA
37
USD
76610-192
76610-194
2-Chloro-5-fluoronicotinic acid 96%
2-Chloro-5-fluoronicotinic acid
Formula:
C₆H₃ClFNO₂ MW: 175.5449 g/mol Melting Pt: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD03092932 CAS Number: 38186-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...