5-Bromo-2,4-difluorobenzaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

PC52118-5G PC52118-25G PC52118-1G
76600-316EA 48 USD
76600-316 76600-314 76600-312
5-Bromo-2,4-difluorobenzaldehyde
5-Bromo-2,4-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 220.9989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842439
CAS Number: 473416-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR