1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Hexafluoroisopropyl methacrylate

PC4756-250G PC4756-100G PC4756-50G
76626-534EA 102 USD
76626-534 76626-530 76626-532
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 98%
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate

Affords low refractive index polymer.

Formula: C₇H₆F₆O₂
MW: 236.12
Boiling Pt: 99 °C
Density: 1.302 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00040105
CAS Number: 3063-94-3
EINECS: 221-309-9
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR